Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant GMCSF (from Bacteria)
Human recombinant GMCSF (from Bacteria)
Nome del fornitore:  Biorbyt
Certificati
Human recombinant GMCSF (from Bacteria)
Nome del fornitore:  Biorbyt

Specifiche

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Bacteria
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biological activity:
    The ED50 was determined by the M1 cell differentiation assay is 0.01 ng/ml, corresponding to specific activity of 100MIU/mg.
  • Protein synonyms:
    GranulocyteMacrophage ColonyStimulating Factor
  • UniProtKB:
    P04141 (Human)
  • Protein/peptide name:
    GMCSF
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MAPARSPSPSTQPWEHVNAIQEARRLLNLSRDTAAEMNETVEVISEMFDLQEPTCLQTRLELYKQ GLRGSLTKLKGPLTMMASHYKQHCPPTPETSCATQIITFESFKENLKDFLLVIPFDCWEPVQE
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 5% Mannitol, pH 7.2.
  • Tested applications:
    SDS-PAGE , FuncS

Specifiche

Informazioni correlate

Frequently Bought Together