Human recombinant FGF acidic (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Biological activity:ED50 is less than 0.5 ng/ml as determined by the dosedependent stimulation of thymidine uptake by 3T3 cells in the presence of Heparin.
Specific Activity of 2.0 x 106 IU/mg. - Protein synonyms:Fibroblast Growth Factor 1
- UniProtKB:P61148 (Mouse)
- Protein/peptide name:FGF acidic
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:FNLPLGNYKKPKLLYCSNGGHFLRILPDGTVDGTRDRSDQHIQLQLSAESAGEVYIKGTETGQYL AMDTEGLLYGSQTPNEECLFLERLEENHYNTYTSKKHAEKNWFVGLKKNGSCKRGPRTHYGQKAI LFLPLPVSSD
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 500mM NaCl, pH 6.6.
- Tested applications:SDS-PAGE , FuncS
Frequently Bought Together