Human recombinant ZNF100 (fromE. coli)
Supplier: Biorbyt
Marketsource
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Zinc Finger 100
- UniProtKB:Q8IYN0 (Human)
- Protein/peptide name:ZNF100
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MGSSHHHHHHSSGLVPRGSHMRHEMVAKPPVICSHFPQDLWAEQDIKDSFQEAILKKYGKYGHDN LQLQKGCKSVDECKVHKEHDNKLNQCLITTQSNIFQCDPSAKVFHTFSNSNRHKIRHTRKKPFK
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 200mM NaCl, 50mM Imidazole, 1mM ZnCl2, 30% Glycerol, pH 8.0.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together