Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant PAFAH beta (from E. coli)
Human recombinant PAFAH beta (from E. coli)
Nome del fornitore:  Biorbyt
Certificati
Human recombinant PAFAH beta (from E. coli)
Nome del fornitore:  Biorbyt

Specifiche

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    PlateletActivating Factor Acetylhydrolase IB Subunit Beta
  • UniProtKB:
    P68402 (Human)
  • Protein/peptide name:
    PAFAH beta
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    SQGDSNPAAIPHAAEDIQGDDRWMSQHNRFVLDCKDKEPDVLFVGDSMVQLMQQYEIWRELFSPL HALNFGIGGDTTRHVLWRLKNGELENIKPKVIVVWVGTNNHENTAEEVAGGIEAIVQLINTRQPQ AKIIVLGLLPRGEKPNPLRQKNAKVNQLLKVSLPKLANVQLLDTDGGFVHSDGAISCHDMFDFLH LTGGGYAKICKPLHELIMQLLEETPEEKQTTIAVEHHHHHH
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris, 150mM NaCl, pH 8.0.
  • Tested applications:
    SDS-PAGE

Specifiche

Informazioni correlate

Frequently Bought Together