Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant PTH 184 (from E. coli)
 :  Biorbyt
 
Human recombinant PTH 184 (from E. coli)
 :  Biorbyt

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biological activity:
    Recombinant PTH is fully biologically active when compared to standards.
    Specific Activity of 1.0 x 104 IU/mg as calculated by the UMR106 cell/cAMP method.
  • Protein synonyms:
    Parathyrin|PTH1 receptor|PTHY_protein.|PTH1|Parathyroid hormone 1|Parathormone|hPTH|PTHR1|PTHR|PTH|Parathyroid hormone|PTH1R
  • UniProtKB:
    P01270 (Human)
  • Protein/peptide name:
    PTH 184
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    SVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNFVALGAPLAPRDAGSQRPRKKEDNVLVESHEK SLGEADKADVNVLTKAKSQ
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 10mM HAc-NaAc, 150mM NaCl, 5% Mannitol, pH 4.0.
  • Tested applications:
    SDS-PAGE , FuncS

 

 

Frequently Bought Together