Human recombinant MDH1 (from E. coli)
Anbieter: Biorbyt
Zertifikate
Über diesen Artikel
Technische Daten
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Malate Dehydrogenase Cytoplasmic
- UniProtKB:P40925 (Human)
- Protein/peptide name:MDH1
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:SEPIRVLVTGAAGQIAYSLLYSIGNGSVFGKDQPIILVLLDITPMMGVLDGVLMELQDCALPLLK DVIATDKEDVAFKDLDVAILVGSMPRREGMERKDLLKANVKIFKSQGAALDKYAKKSVKVIVVGN PANTNCLTASKSAPSIPKENFSCLTRLDHNRAKAQIALKLGVTANDVKNVIIWGNHSSTQYPDVN HAKVKLQGKEVGVYEALKDDSWLKGEFVTTVQQRGAAVIKARKLSSAMSAAKAICDHVRDIWFGT PEGEFVSMGVISDGNSYGVPDDLLYSFPVVIKNKTWKFVEGLPINDFSREKMDLTAKELTEEKES AFEFLSSALEHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris, 150mM NaCl, pH 8.0.
- Tested applications:SDS-PAGE
Technische Daten
Frequently Bought Together