Human recombinant HUS1 (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Checkpoint HUS1
- UniProtKB:O60921 (Human)
- Protein/peptide name:HUS1
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MGSSHHHHHHSSGLVPRGSHMKFRAKIVDGACLNHFTRISNMIAKLAKTCTLRISPDKLNFILCD KLANGGVSMWCELEQENFFNEFQMEGVSAENNEIYLELTSENLSRALKTAQNARALKIKLTNKHF PCLTVSVELLSMSSSSRIVTHDIPIKVIPRKLWKDLQEPVVPDPDVSIYLPVLKTMKSVVEKMKN ISNHLVIEANLDGELNLKIETELVCVTTHFKDLGNPPLASESTHEDRNVEHMAEVHIDIRKLLQF LAGQQVNPTKALCNIVNNKMVHFDLLHEDVSLQYFIPALSVEHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 40% Glycerol, pH 8.0 .
- Tested applications:SDS-PAGE
Frequently Bought Together