Human recombinant CD47 (from HEK293 cells)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Leukocyte Surface Antigen CD47
- UniProtKB:Q08722 (Human)
- Protein/peptide name:CD47
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:QLLFNKTKSVEFTFCNDTVVIPCFVTNMEAQNTTEVYVKWKFKGRDIYTFDGALNKSTVPTDFSS AKIEVSQLLKGDASLKMDKSDAVSHTGNYTCEVTELTREGETIIELKYRVVSWFSPVDHHHHHH
- Formulation:Lyophilized from a 0.2 um filtered solution of 10mM Tris-Citrate, 150mM NaCl, pH 8.0.
- Tested applications:SDS-PAGE
Frequently Bought Together