Human recombinant ARSA (from HEK293 cells)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Arylsulfatase A
- UniProtKB:P15289 (Human)
- Protein/peptide name:ARSA
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:RPPNIVLIFADDLGYGDLGCYGHPSSTTPNLDQLAAGGLRFTDFYVPVSLCTPSRAALLTGRLPV RMGMYPGVLVPSSRGGLPLEEVTVAEVLAARGYLTGMAGKWHLGVGPEGAFLPPHQGFHRFLGIP YSHDQGPCQNLTCFPPATPCDGGCDQGLVPIPLLANLSVEAQPPWLPGLEARYMAFAHDLMADAQ RQDRPFFLYYASHHTHYPQFSGQSFAERSGRGPFGDSLMELDAAVGTLMTAIGDLGLLEETLVIF TADNGPETMRMSRGGCSGLLRCGKGTTYEGGVREPALAFWPGHIAPGVTHELASSLDLLPTLAAL AGAPLPNVTLDGFDLSPLLLGTGKSPRQSLFFYPSYPDEVRGVFAVRSGKYKAHFFTQGSAHSDT TADPACHASSSLTAHEPPLLYDLSKDPGENYNLLGGVAGATPEVLQALKQLQLLKAQLDAAVTFG PSQVARGEDPALQICCHPGCTPRPACCHCPDPHAVDHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 7.5.
- Tested applications:SDS-PAGE
Frequently Bought Together