Human recombinant Annexin A2 (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:PAP-IV|p36|Lipocortin II|Protein I|LPC2D|Placental anticoagulant protein IV|Calpactin I heavy chain|CAL1H|ANX2L4|Calpactin-1 heavy chain|Annexin II|ANX2|Chromobindin-8|Annexin-2
- UniProtKB:P07355 (Human)
- Protein/peptide name:Annexin A2
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MSTVHEILCKLSLEGDHSTPPSAYGSVKAYTNFDAERDALNIETAIKTKGVDEVTIVNILTNRSN AQRQDIAFAYQRRTKKELASALKSALSGHLETVILGLLKTPAQYDASELKASMKGLGTDEDSLIE IICSRTNQELQEINRVYKEMYKTDLEKDIISDTSGDFRKLMVALAKGRRAEDGSVIDYELIDQDA RDLYDAGVKRKGTDVPKWISIMTERSVPHLQKVFDRYKSYSPYDMLESIRKEVKGDLENAFLNLV QCIQNKPLYFADRLYDSMKGKGTRDKVLIRIMVSRSEVDMLKIRSEFKRKYGKSLYYYIQQDTKG DYQKALLYLCGGDD
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 1mM EDTA, pH 7.5 .
- Tested applications:SDS-PAGE
Frequently Bought Together