MCSF
Codice catalogo BIRBORB97818-10
Nome del fornitore: Biorbyt
Numero CAS:
Specifiche
- Pk:10 µG
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 5 ng/ml, corresponding to a specific activity of 2x105 IU/mg, calculated by the dose dependent stimulation of the proliferation of murine M-NFS-60indicator cells.
- UniProtKB:P09603
- Protein/peptide name:MCSF
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized M-CSFin sterile bidest water not less than 100 g/ml. For longterm storage it is recommended to add a carrier protein(0.1% HSA or BSA).
- Molecular weight:37 kDa
- Formulation:Lyophilized