GH-20K
BIRBORB97769-100
: Biorbyt
:
- Pk:100 µG
- Storage conditions:Store at−20 °C.
- Biological activity:3 U/mg
- UniProtKB:P01241
- Protein/peptide name:GH-20K
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute and dilute the lyophilizedPlacental-GH in 0.4% NaHCO3 adjusted to pH 10.0,which can then be further diluted to other aqueoussolutions. It is recommended when diluting to below 10ug/ml that the dilution solution will conta
- Molecular weight:20 kDa
- Formulation:Lyophilized. Pituitary Growth Hormone protein waslyophilized from a 1 mg/ml solution with 0.0045 mMNaHCO3 and pH 10.0.