Rat recombinant IL6 (from E. coli)
Catalog # BIRBORB82020-10
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Protein synonyms:Interleukin BSF 2
- Protein/peptide name:IL6
- Purity:Purified.
- Formulation:Lyophilised.