Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant TNF alpha (from E. coli)
Human recombinant TNF alpha (from E. coli)
Catalog # BIRBORB82061-50
Supplier:  Biorbyt
CAS Number:  
Human recombinant TNF alpha (from E. coli)
Catalog # BIRBORB82061-50
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    APC1
  • Protein/peptide name:
    TNF alpha
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.

Specifications

About this item