Human recombinant IL1 alpha (from E. coli)
Catalog # BIRBORB82046-10
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:Hematopoietin 1
- Protein/peptide name:IL1 alpha
- Purity:Purified.
- Formulation:Lyophilised.