Order Entry
Switzerland
ContactUsLinkComponent
Human insulin like growth factor binding protein-1 (IGFBP-1), calibrator grade (from amniotic fluid)
Human insulin like growth factor binding protein-1 (IGFBP-1), calibrator grade (from amniotic fluid)
Catalog # BIRBORB81872-100
Supplier:  Biorbyt
CAS Number:  
Human insulin like growth factor binding protein-1 (IGFBP-1), calibrator grade (from amniotic fluid)
Catalog # BIRBORB81872-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Source:
    Amniotic fluid
  • Species:
    Human
  • Protein synonyms:
    Pregnancy-Associated Plasma-1
  • Protein/peptide name:
    Insulin Like Growth Factor Binding Protein-1 (IGFBP-1), Calibrator Grade
  • Purity:
    Calibrator.
  • Formulation:
    Lyophilised.
  • Tested applications:
    ELISA

Specifications

About this item