Lipoprotein, hgh density (from Plasma)
Catalog # BIRBORB82282-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:Plasma
- Protein synonyms:Lipoprotein
- Protein/peptide name:Lipoprotein, High Density
- Purity:Purified.
- Formulation:Preservative: 1 mm Sodium Azide