Order Entry
Switzerland
ContactUsLinkComponent
 
Toxoplasma gondii recombinant Toxoplasma gondii MIC 3 protein (from Escherichia coli)
  BIRBORB82491-1
 :  Biorbyt
 :  
Toxoplasma gondii recombinant Toxoplasma gondii MIC 3 protein (from Escherichia coli)
  BIRBORB82491-1
 :  Biorbyt
 :  

 

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Toxoplasma gondii
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein synonyms:
    Toxoplasma gondii MIC 3
  • Protein/peptide name:
    Toxoplasma gondii MIC 3 Protein
  • Purity:
    GS-4B Sepharose-affinity Purified.
  • Formulation:
    Liquid, 50 mM Tris-HCl, 1.5M Urea containing 50% Glycerol
  • Tested applications:
    ELISA , WB