Recombinant Hherpes simplex virus-2 (HSV-2) glycoprotein G (from E. coli)
Catalog # BIRBORB82468-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein/peptide name:Herpes Simplex Virus-2 (HSV-2) Glycoprotein G
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB