Order Entry
Switzerland
ContactUsLinkComponent
 
Mouse VEGF (121) (from E. coli)
 :  Biorbyt
 
Mouse VEGF (121) (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Vascular endothelial growth factor A
  • Protein/peptide name:
    VEGF (121)
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized VEGF-121 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKRPR R.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution with no additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

 

 

Frequently Bought Together