Human recombinant PEDF (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Gene ID:5176
- Protein synonyms:EPC 1
- Protein/peptide name:PEDF
- Purity:> 95% as determined by SDS-PAGE.
- Solubility:Add deionized water to working concentration of 0.5mg/ml and let the lyophilized pellet dissolve completely.
- Sequence:MQNPASPPEEGS PDPDSTGALVEEE DPFFKVPVNKLA AAVSNFGYDLYR VRSSMSPTTNVL LSPLSVATALSALS LGAEQRTESIIHRA LYYDLISSPDIHGT YKELLDTVTAPQK NLKSASRIVFEKK LRIKSSFVAPLEKS YGTRPRVLTGNPR LDLQEINNWVQA QMKGKLARSTKE IPDEISILLLGVAHF KGQWVTKFDSRK TSLEDFYLDEERTV RVPMMSDPKAVLRY GLDSDLSCKIAQLPL TGSMSIIFFLPLKVTQ NLTLIEESLTSEFIH DIDRELKTVQAVLTVP KLLSYEGEVTKS LQEMKLQSLFD SPDFSKITGKPI KLTQVEHRAGF EWNEDGAGTTPSPG LQPAHLTFPL DYHLNQPFIF VLRDTDTGALLFIG KILDPRGP.
- Formulation:The sterile filtered concentrated (1mg/ml) protein solution was lyophilized with 20 mM sodium phosphate buffer 150mM NaCl pH-7.4.
- Tested applications:SDS-PAGE
Frequently Bought Together