Human recombinant MIP1B (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The Activity is calculated by the ability to chemoattract Human blood monocytes using concentration of 5-20 ng/ml.
- Protein synonyms:Small inducible cytokine A4
- Protein/peptide name:MIP1B
- Purity:> 99.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Macrophage Inflammatory Protein-1b in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be, Ala-Pro-Met-Gly-Ser.
- Formulation:Lyophilized from concentrated (1 mg/ml) solution in water containing no additives.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together