Bovine FGF basic (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Bovine
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50, measured in mitogenic assay using quiescent NR6R-3T3 fibroblasts was found to be <0.1 ng/ml, corresponding to specific activity of 3,000,000 Units/mg.
- Protein synonyms:HBGH-2
- Protein/peptide name:FGF basic
- Purity:> 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Fibroblast Growth Factor-2 Bovine Recombinant in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Met-Ala-Ala-Gly-Ser.
- Formulation:The FGF-b Bovine was lyophilized from concentrated (1mg/ml) sterile solution containing 1%HSA.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together