Human fibronectin (from plasma)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Plasma
- Species:Human
- Storage conditions:Lyophilized Fibronectin although stable at room temperature for week, should be stored desiccated below −18 °C. Upon reconstitution Fibronectin should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
- Protein synonyms:CIG
- Protein/peptide name:Fibronectin
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Solubility:Purified fibronectin has tendency to form insoluble aggregates upon reconstitution. We suggest to reconstittue the 1mg Fibronectin with chaotropic agent such as urea at room temperature at concentration of 0.2mg/ml using sterile water. Let stand 1-2 hours
- Formulation:The Fibronectin was lyophilized from concentrated 1mg/ml solution containing 0.05M Tris HCl pH-7.5 and 0.1M NaCl.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together