Mouse recombinant FGF 9
: Biorbyt
- Protein/peptide type:Recombinant
- Species:Mouse
- Storage conditions:Store at −20 °C.
- Biological activity:The ED50 as determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 106 units/mg.
- Protein synonyms:Growth Factor-9
- UniProtKB:P54130
- Protein/peptide name:FGF 9
- Purity:>95%
- Solubility:It is recommended to reconstitute the lyophilized Fibroblast Growth Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:PLGEVGSYFG VQDAVPFGNV PVLPVDSPVL LNDHLGQSEA GGLPRGPAVT DLDHLKGILR RRQLYCRTGF HLEIFPNGTI QGTRKDHSRF GILEFISIAV GLVSIRGVDS GLYLGMNEKG ELYGSEKLTQ ECVFREQFEE NWYNTYSSNL YKHVDTGRRY YVALNKDGTP REGTRTKRHQ KFTHFLPRPV DPDKVPELYK DILSQS
- Formulation:The protein was lyophilized from 10mM Tris, pH 8.0, 0.15M Amonium Sulfate.
- Tested applications:SDS-PAGE
Frequently Bought Together