Mouse recombinant RELM alpha
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Storage conditions:Store at −20 °C.
- Biological activity:Not Available at this time.
- Gene ID:57262
- Protein synonyms:Resistin-like alpha
- UniProtKB:Q9EP95
- Protein/peptide name:RELM alpha
- Purity:>98%
- Solubility:At 0.1mg/ml of deionized sterile water.
- Sequence:MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
- Formulation:Sterile filtered and lyophilized from 0.5mg/ml in 10mM sodium phosphate buffer, pH 7.5.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together