Complement, rabbit serum
Rabit serum complement, Lyophilised
Catalog # 234400-5
Supplier: Merck Millipore (Oncogene)
Jump to:
Product Details & Documents
Native, rabbit serum complement is tested for suitability for HLA-ABC serology. Useful as an active source of complement for use with hybridoma antibodies, alloantisera, and related applications in which a non-cytotoxic source is required. Total lympholytic complement activity determined under conditions of NIH two-step cytotoxicity test that uses defined HLA isoantisera.
- Activity present at a titer of at least 1:4
- No intrinsic cytotoxicity against human peripheral blood lymphocytes
- Intended for micro and macro lymphocyte cytotoxicity testing with human or animal sera
Specifications
- Serum format:Lyophilised
- Description:Rabit serum complement