Buffer, PBS (1X, Dulbecco's) powder pack
Powder mixture to prepare PBS buffer (1X) according to Dulbecco.
Composed of di-sodium hydrogen phosphate anhydrous (1150 mg/l), potassium chloride (200 mg/l), potassium dihydrogen phosphate (200 mg/l) and sodium chloride (8000 mg/l).
Powder mixture to prepare PBS buffer (1X) according to Dulbecco.
Composed of di-sodium hydrogen phosphate anhydrous (1150 mg/l), potassium chloride (200 mg/l), potassium dihydrogen phosphate (200 mg/l) and sodium chloride (8000 mg/l).
Product Family Options
- DescriptionPk
- PBS buffer (1X, Dulbecco's), 10 l1 KIT1 * 1 KITSFr. 48.40
- PBS buffer (1X, Dulbecco's), 100 l1 KIT1 * 1 KITSFr. 140.00