Packed GC columns, 2abc refinery column set
Catalog # REST88000-810
Supplier: Restek
Specifications
- Description:Analytical controls configuration
- Seq:01
- Pk:1
Specifications
About this item
Special columns for unsaturated light hydrocarbons.
- Faster separations of C1 to C4 hydrocarbons
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.