ContactUsLinkComponent
Ice borer
Ice borer
Catalog # SAMP6005A-500
Supplier:  Sampling Systems
undefined
Ice borer
Catalog # SAMP6005A-500
Supplier:  Sampling Systems
Specifications
  • Ø:
    25 mm
  • Description:
    Ice borer
  • Length:
    500 mm
  • Material:
    Stainless steel 316
  • Pk:
    1
Specifications
About this item
Ice borer specifically designed for ice, wax, and other similar materials.
  • Sharp cutting tip