Hygiene masks, PP
Catalog # 114-9101
Supplier: HUBEI ORIENT INTL TRADING
Specifications
- Type:With earloops
- Colour:Green
- Seq:02
- Environmentally Preferable:
- Pk:50
Specifications
About this item
Hygiene masks made from 3-ply construction of PP and Meltblown PP.
- Easy breathing, non-irritating
- Excellent bacterial and particle filtration
- Full length aluminum nose piece for a comfortable fit
- For industrial and clean room use only; do not use for protection against COVID-19
- This product is not a medical device for medical or clinical use within the meaning of Regulation (EU) 2017/745 / Directive 93/42/EEC
- This product is not a personal protective equipment (PPE) within the meaning of Regulation (EU) 2016/425