ContactUsLinkComponent
Isolation gowns
Isolation gowns
Catalog # HOIT110412
undefined
Isolation gowns
Catalog # HOIT110412
Specifications
  • Anti-static:
    +
  • Closure type:
    Back fastening
  • Garment application:
    Medical,
    Clinical
  • Gender:
    Unisex
  • Colour:
    White
  • Pk:
    100
Specifications
About this item
Full length gown made of soft PP fabric.
  • Elastic cuffs
  • Extra long ties sewn at waist and neckline
  • Medical repellence grade (anti-alcohol, anti-blood, anti-oil), anti-static
  • No dust and particle release