ContactUsLinkComponent
Detergents for glassware washers, ProCare Lab 11 MA
Detergents for glassware washers, ProCare Lab 11 MA
Catalog # MIEL10266720
Supplier:  MIELE
undefined
Detergents for glassware washers, ProCare Lab 11 MA
Catalog # MIEL10266720
Supplier:  MIELE
Specifications
  • Capacity:
    10 kg
  • Description:
    Alkaline detergent powder, ProCare Lab 11 MA
  • Pk:
    10 kg
  • pH range:
    11,7 - 11,9
Specifications
About this item
Powder, mildly alkaline, foam-retardant action detergent for machine based cleaning of laboratory glassware and laboratory utensils.
  • Provides good protection to surfaces
  • Removes blood, protein and food residues
  • Free from oxidative agents
ProCare Lab 11 MA is for general-purpose use in medical, biological and chemical laboratories.
Note: Suitable for stainless steel, laboratory glassware, ceramics, plastics such as PE, PP, PVDF, PTFE. Conditionally suitable for use on non ferrous metals and aluminium. Not suitable for anodised aluminium.