ContactUsLinkComponent
Portable refractometer for automotive care, Multi-Chek
Portable refractometer for automotive care, Multi-Chek
Catalog # 635-0509
Supplier:  REICHERT
undefined
Portable refractometer for automotive care, Multi-Chek
Catalog # 635-0509
Supplier:  REICHERT
Specifications
  • Model:
    Multi-Chek
  • Description:
    Digital refractometer for glycol coolants/ battery electrolytes/windshield fluid tester
  • Seq:
    001
  • Pk:
    1
Specifications
About this item
Three-in-one tester for measuring freezing point of engine coolants with ATC (ethylene glycol and propylene glycol); the specific gravity and condition of battery electrolytes and lastly the freezing point of the wind screen washer fluid.