Glove bag
Catalog # 112-9990
Supplier: INSTRUMENTS FOR RESEARCH
Specifications
- Description:Glove bag, 686×686×381 mm (when inflated)
- Pk:1
Specifications
About this item
PE glove bag with inflatable glove chamber, Handy-Lok closure, turned in gloves and one equipment opening.
- Greater flexibility than a rigid glove box
- Excels for working with air sensitive or moisture sensitive materials
- Economical enough to use once and discard or durable enough to use over and over
- Can be purged quickly and with less gas
Delivery: Supplied with one pair of cotton gloves.