Safety storage cabinets, Type 30, K-PHOENIX-30
Catalog # MSGA30343001
Supplier: asecos
Specifications
- Bezeichnung:Hazardous substance center, model K30.197.120.FWAS
Body light grey RAL 7035 / doors light grey RAL 7035 - B×T×H:1202×599×1968 mm
- VE:1
- Gewicht:330 kg
- Innen B×T×H:532×519×1691 mm
Specifications
About this item
Safe and approved storage of aggressive and flammable hazardous substances in working areas.
- Robust construction and longevity - triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one - 2 compartments (vertical separation) allow the storage of flammable liquids and aggressive hazardous materials in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use - doors lockable with profi le cylinder (integration in an existing locking system possible) / cylinder lock
Compartment for the storage of aggressive, non flammable hazardous materials, metal-free tray shelves and collecting sumps.
Ordering information: Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check the website or contact Avantor for further information.
Delivery: Delivery prices available upon request.