Overalls, 4500
Supplier: 3M
Certificates
About this item
This simple overall made of durable PP fabric is suitable for non hazardous applications where hygiene and cleanliness are important.
- Excellent economic solution to keep the wearer clean or dry
- Breathable and light duty protection
- Two-way zip with storm flap
- Elasticated 2-panel hood, cuffs, ankles and waist
Typical applications include: Domestic DIY use, light duty industrial cleaning and maintenance applications.
Gewicht: 45 g/m²
Certifications: Cat. I
Specifications
- Catalog No:
- MMMA4500W3XL
- MMMA4500W2XL
- MMMA4500WL
- MMMA4500W4XL
- MMMA4500WS
- MMMA4500WXL
- MMMA4500WM
- Modell:
- 4500
- 4500
- 4500
- 4500
- 4500
- 4500
- 4500
- Stoff:
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- Anwendungsbereich Outfit:
- Allgemeine Anwendungen, Labor, Industrie
- Allgemeine Anwendungen, Labor, Industrie
- Allgemeine Anwendungen, Labor, Industrie
- Allgemeine Anwendungen, Labor, Industrie
- Allgemeine Anwendungen, Labor, Industrie
- Allgemeine Anwendungen, Labor, Industrie
- Allgemeine Anwendungen, Labor, Industrie
- Gewicht:
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²