Nitrile gloves, uvex u-fit lite
Supplier: UVEX
Certificates
About this item
These disposable safety gloves provide reliable spray protection when handling chemicals such as acids, alkalis, solids or aqueous saline solutions.
- Good grip thanks to roughened fingertips
- Good mechanical resistance
- Silicone-free according to imprint test, accelerator-free
- Outstanding tactile feel
- Very good fit, extremely high flexibility
- Highly resistant to grease and oil
Typical applications include: Precision assembly work, inspection, short periods handling chemicals, paint shop (as splash protection), food processing, and gentle cleaning.
Certifications: EN ISO 374-1 Type C (K); AQL 1,5
Food contact approved
Food contact approved
Specifications
- Catalog No:
- UVEX6059709
- UVEX6059707
- UVEX6059708
- Model:
- lite
- lite
- lite
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- EN norm:
- EN ISO 374-1 Type C (K)
- EN ISO 374-1 Type C (K)
- EN ISO 374-1 Type C (K)
- Food contact approved:
- +
- +
- +