ContactUsLinkComponent
Qualitative filter papers, grade 598, Whatman™
Qualitative filter papers, grade 598, Whatman™
Certificates
undefined
Qualitative filter papers, grade 598, Whatman™
Specifications
  • Catalog No:
  • 515-8055
  • 515-4202
  • WHAT10312000
  • Grade:
  • 598
  • 598
  • 598
  • Weight:
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • Thickness:
  • 320 µm
  • 320 µm
  • 320 µm
  • Particle retention:
  • 10 µm
  • 10 µm
  • 10 µm
  • Filtration speed:
  • 50 s (Herzberg)
  • 50 s (Herzberg)
  • 50 s (Herzberg)