ContactUsLinkComponent
Rack for sedimentation plates
Rack for sedimentation plates
Catalog # HEIP1465190001
Supplier:  MERCK LIFE SCIENCE
Rack for sedimentation plates
Catalog # HEIP1465190001
Supplier:  MERCK LIFE SCIENCE
Specifications
  • Description:
    Rack, stainless steel, 240 mm, for 10× Ø 90 mm settle plates
  • Pk:
    1
Specifications
About this item
High-quality stainless steel sedimentation plate rack suitable for isolators and cleanroom applications (resistant against H₂O₂ and disinfectants).
  • Locking device aids safe, convenient handling
  • Autoclavable