Cell culture storage bottles, polystyrene, Corning®
Supplier: Corning
Certificates
About this item
Disposable polystyrene bottles for storage of media, buffers and other aqueous solutions.
- Low profile, easy grip style has sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimise the risk of contamination
- Bottles can be used with Corning® vacuum filter systems
- Sterile, certified non-pyrogenic
Specifications
- Catalog No:
- 700-1786
- 734-1847
- 734-1897
- 734-1825
- 734-1824
- 700-1783
- 700-1787
- 700-1785
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta