Reading magnifiers, easyPOCKET
Supplier: ESCHENBACH OPTIK
Certificates
About this item
These pocket magnifiers provide magnification and illumination in a credit card format.
- Ultra-flat and lightweight design
- Diffractive aspheric PXM® lightweight lens
- SMD LED illumination, which is switched on when you slide the lens out of its case
- Easy handling - No bulb change required
Delivery information: Supplied with two button cells.
Frequently Bought Together