VWR®, UV/Visible spectroscopy wavelength standards
Supplier: VWR Chemicals
Certificates
About this item
Wavelength standards (certified at 0,1; 0,2; 0,5; 1 and 2 nm slit widths) packed in six permanently sealed UV cuvettes or 100 ml amber bottles.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together