ALLN
10 mg
Katalognr. 786-057
Anbieter: G-Biosciences
Direkt zu
Product Details & Documents
ALLN is a cell permeable peptide aldehyde inhibitor of calpain I and to a lesser extent calpain II. It also inhibits other neutral cysteine proteases, cathepsin B and L and the proteasome.
Technische Daten
Technische Daten
- Pk:10 mg
- Cat. No.:786-057
- Protein/peptide name:ALLN
Technische Daten