Order Entry
Switzerland
ContactUsLinkComponent
VWR® PESTINORM®, Methanol, Secondary Reference Standard for GC
VWR® PESTINORM®, Methanol, Secondary Reference Standard for GC
Catalog # 85670.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® PESTINORM®, Methanol, Secondary Reference Standard for GC
Catalog # 85670.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Methanol standard for GC

Specifications

About this item

A secondary GC solvent reference standard for various chromatographic and analytical applications.

  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID