Recombinant thioredoxin (from E. coli), HIS tag
Catalog # ENZOADISPP8921
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Seq:01
- Tag sequence:HIS Tag
- Storage conditions:–80 °C
- Protein synonyms:Trx
- Protein/peptide name:Thioredoxin
- Purity:>95%
- Formulation:Liquid
- Shipping temperature:Shipped on dry ice
Specifications
About this item
Thioredoxin (Trx) is a redox protein of approximately 12 kDa. Its primary domain is conserved across a number of Trx family members and contains a conserved catalytic site Cys-Gly-Pro-Cys. It is ubiquitous and found in many organisms from bacteria to mammals. Trx has been shown to function in cell proliferation, redox signaling and inhibition of apoptosis.
- Produced from E. coli
- E. coli thioredoxin is fused at the N-terminus to a His-tag
- Purified by multi-step chromatography
- Supplied in 50 mM TRIS, pH 7,5, containing 100 mM sodium chloride and 1 mM EDTA