Order Entry
Switzerland
ContactUsLinkComponent
ELISA kits, LVV hemorphin 7
ELISA kits, LVV hemorphin 7
Catalog # ENZOADI900205
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
ELISA kits, LVV hemorphin 7
Catalog # ENZOADI900205
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Description:
    LVV Hemorphin 7 ELISA kit, 96 wells
  • Seq:
    02
  • Sensitivity:
    6,1 pg/ml (range 9,8 - 10000 pg/ml)

Specifications

About this item

The LVV Hemorphin 7 ELISA Kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent LVV Hemorphin 7 in serum and tissue homogenate samples.

  • Sensitive - reliably measure as little as 6,1 pg/ml of LVV Hemorphin 7
  • Rapid - results in just 3 hours from up to 40 samples in duplicate
  • Specific - low cross reactivity to similar peptides
  • Easy to use liquid colour coded reagents reduce errors

For the quantitative determination of species independent LVV Hemorphin 7 in serum and tissue homogenate samples.

The LVV Hemorphin 7 EIA kit is a colorimetric, competitive immunoassay kit with results in just 3 hours.

Ordering information: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Delivery: The kit contains microtiter plate, conjugate, antibody, assay buffer 16, wash buffer concentrate, standard, TMB substrate, stop solution 2, and Streptavidin-HRP.