Angiotensin II ELISA Kits
Catalog # ENZOADI900204
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Target protein:Angiotensin II
- Description:Angiotensin II ELISA kit, 96 wells
- Seq:02
Specifications
About this item
The Angiotensin II ELISA Kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent Angiotensin II in plasma and serum samples.
- Results in 3,5 hours
- Highly sensitive kit detecting as little as 4,6 pg/ml
- Highly specific with low cross-reactivity to similar peptides
- Fully quantitative results that surpass semi-quantitative western blot analysis
- Easy to use liquid colour coded reagents reduce errors
- Ready to use reagents and pre-coated plates reduce errors and save your time
Cross reactivity: Angiotensin II (100%), Angiotensin A (100%), Angiotensin III (100%), Angiotensin IV (100%), Angiotensin I (0,319%), Angiotensin (1-9) (0,103%), LVV-hemorphin 7 (0,067%), Angiotensin (1-7) (0,053%), Bradykinin (0,022%).
Delivery: Kit contains microtiter plate, conjugate, antibody, Assay Buffer 16, wash buffer concentrate, standard, TMB substrate, Stop Solution 2, and Streptavidin-HRP.