Order Entry
Switzerland
ContactUsLinkComponent
Carrez II solution potassium hexacyanoferrate(II) 0.25 mol/l in aqueous solution
Environment
Carrez II solution potassium hexacyanoferrate(II) 0.25 mol/l in aqueous solution
Catalog # 85733.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Environment
Carrez II solution potassium hexacyanoferrate(II) 0.25 mol/l in aqueous solution
Catalog # 85733.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer:
    0,2375 - 0,2625 mol/l

Specification Test Results

About this item

 :
Density:1.060 g/cm³ (20 °C)
EINECS:237-722-2
 :GHS09
Storage Temperature:Ambient