Hydrochloric acid 6 mol/l (6 N) for biopharmaceutical production
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.09 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
:GHS07
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer:5.7 - 6.3 mol/l
- Appearance:Passes test
- Free bromine or chlorine:Passes test
- Identification:Passes test
- Colouration:Max. 10 APHA
- Evaporation residue:Max. 100 ppm
- Free chlorine:Max. 1 ppm
- Heavy metals (as Pb):Max. 1 ppm
- Ignition residue (SO4):Max. 80 ppm
- SO3 (Sulphite):Max. 10 ppm
- SO4 (Sulphate):Max. 5 ppm
- As (Arsenic):Max. 1.5 ppm
- Residual metal catalysts:Unlikely to be present
- Residual solvents (ICH Q3C):Excl.by manufact. process
- Endotoxin:Max. 0.5 EU/ml
- Filtered at 0.2 µm:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number28579898.pdf spec_VWRC85401.320 spec_VWRC85401.460
Product specification
85401_HYDROCHLORIC ACID 6M BIOPHARMA_BSE_REV0
BSE/TSE Certificates
Frequently Bought Together